Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-193 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-193 Supplier Abnova Corporation Supplier No. PAB0382
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VDAC2.

Sequence: KKSGKIKSSYKRE

Specifications

Antigen VDAC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VDAC2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene VDAC2
Gene Alias FLJ23841
Gene Symbols VDAC2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 120-132 of human VDAC2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7417
Target Species Bovine, Canine, Chimpanzee, Feline, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.