Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFA, Rabbit, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117438 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-438 Supplier Abnova Corporation Supplier No. PAB15095
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VEGFA.

This gene is a member of the PDGF/VEGF growth factor family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Elevated levels of this protein is linked to POEMS syndrome, also known as Crow-Fukase syndrome. Mutations in this gene have been associated with proliferative and nonproliferative diabetic retinopathy. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. There is also evidence for the use of non-AUG (CUG) translation initiation sites upstream of, and in-frame with the first AUG, leading to additional isoforms. [provided by RefSeq

Specifications

Antigen VEGFA
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of VEGFA.
Dilution Western Blot (1 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene VEGFA
Gene Alias MGC70609/VEGF/VEGF-A/VPF
Gene Symbols VEGFA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 150-250 of human VEGFA.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity This is specific to VEGFA. This antibody is useful for Western blot analysis, where a band is seen at ∼42 KDa with a chimera transfected lysate, representing the homodimer VEGFA. Another faint band is seen at ∼21 KDa, representing the monodomer form of VEGFA.
Target Species Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.