Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFA Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-723 Supplier Abnova Corporation Supplier No. PAB13526

Rabbit polyclonal antibody raised against recombinant Vegfa.

Specifications

Antigen VEGFA
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Vegfa.
Dilution ELISA (0.5-5 ug/mL) Western Blot (0.1-0.2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Gene Vegfa
Gene Alias Vegf/Vegf-a/Vegf120/Vegf164/Vegf188/Vpf
Gene Symbols Vegfa
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Vegfa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22339
Test Specificity This antibody reacts with VEGFA isoforms.
Target Species Human, Mouse
Content And Storage Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.