Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIL1, Rabbit, Clone: SP145, Abnova™

Catalog No. 89109256 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-256 Supplier Abnova Corporation Supplier No. MAB11042

Rabbit monoclonal antibody raised against synthetic peptide of VIL1.

This gene encodes a member of a family of calcium-regulated actin-binding proteins. This protein represents a dominant part of the brush border cytoskeleton which functions in the capping, severing, and bundling of actin filaments. Two mRNAs of 2.7kb and 3.5kb have been observed; they result from utilization of alternate poly-adenylation signals present in the terminal exon. [provided by RefSeq

Specifications

Antigen VIL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP145
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of VIL1.
Dilution Western Blot (1:100) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene VIL1
Gene Alias D2S1471/VIL
Gene Symbols VIL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of VIL1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.