Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIPR2 Rabbit anti-Bovine, Human, Monkey, Polyclonal Antibody, Abnova™

Catalog No. 89129239 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-239 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-239 Supplier Abnova Corporation Supplier No. PAB27722
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VIPR2.

Vasoactive intestinal peptide (VIP; MIM 192320) and pituitary adenylate cyclase activating polypeptide (PACAP; MIM 102980) are homologous peptides that function as neurotransmitters and neuroendocrine hormones. While the receptors for VIP and PACAP share homology, they differ in their substrate specificities and expression patterns. See VIPR1 (MIM 192321) and ADCYAP1R1(MIM 102981).[supplied by OMIM

Specifications

Antigen VIPR2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VIPR2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene VIPR2
Gene Alias FLJ16511/VPAC2/VPCAP2R
Gene Symbols VIPR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acid at internal region of human VIPR2.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7434
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Bovine, Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.