Learn More
VISA Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VISA.
Supplier: Abnova Corporation PAB13251
Description
Double-stranded RNA viruses are recognized in a cell type-dependent manner by the transmembrane receptor TLR3 (MIM 603029) or by the cytoplasmic RNA helicases MDA5 (MIM 606951) and RIGI (ROBO3; MIM 608630). These interactions initiate signaling pathways that differ in their initial steps but converge in the activation of the protein kinases IKKA (CHUK; MIM 600664) and IKKB (IKBKB; MIM 603258), which activate NFKB (see MIM 164011), or TBK1 (MIM 604834) and IKKE (IKBKE; MIM 605048), which activate IRF3 (MIM 603734). Activated IRF3 and NFKB induce transcription of IFNB (IFNB1; MIM 147640). For the TLR3 pathway, the intermediary molecule before the pathways converge is the cytoplasmic protein TRIF (TICAM1; MIM 607601). For RIGI, the intermediary protein is mitochondria-bound IPS1 (Sen and Sarkar, 2005 [PubMed 16239922]).[supplied by OMIM
Specifications
VISA | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of VISA. | |
In PBS (0.02% sodium azide) | |
CARDIF/DKFZp547C224/DKFZp666M015/FLJ27482/FLJ35386/FLJ38051/FLJ41962/IPS-1/IPS1/KIAA1271/MAVS/MGC3260 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
VISA | |
VISA | |
A synthetic peptide corresponding to N-terminus 13 amino acids of human VISA. | |
RUO | |
57506 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only