Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitellogenin (VTG), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-628 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-628 Supplier Abnova Corporation Supplier No. PAB13582
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length native Vitellogenin (VTG).

Specifications

Antigen Vitellogenin (VTG)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length native Vitellogenin (VTG).
Dilution Western Blot (1:5000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from serum
Host Species Rabbit
Immunogen Native purified Senegalese sole Vitellogenin.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species European seabass, Salmonid species, Senegalese sole
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.