Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VN1R1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89129231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-231 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-231 Supplier Abnova Corporation Supplier No. PAB27715
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VN1R1.

Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. [provided by RefSeq

Specifications

Antigen VN1R1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VN1R1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7-18 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene VN1R1
Gene Alias V1RL1/VNR19I1/ZVNH1/ZVNR1
Gene Symbols VN1R1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acid at extracellular domain of human VN1R1.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57191
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except VN1R3 (53%).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.