Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS13A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89122932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-932 Supplier Abnova Corporation Supplier No. PAB21438
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant VPS13A.

The protein encoded by this gene may control steps in the cycling of proteins through the trans-Golgi network to endosomes, lysosomes and the plasma membrane. Mutations in this gene cause the autosomal recessive disorder, chorea-acanthocytosis. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq

Sequence: KCGLVMLNNLVKAFTEAATGSSADFVKDLAPFMILNSLGLTISVSPSDSFSVLNIPMAKSYVLKNGESLSMDYIRTKDNDHFNAMTSLSSKLFFILLTPVNHSTADKIPLTKVGRRLYTVRHRESGVERSIVCQIDTVEGSKKVTIRSPV

Specifications

Antigen VPS13A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant VPS13A.
Dilution Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene VPS13A
Gene Alias CHAC/CHOREIN/FLJ42030/KIAA0986
Gene Symbols VPS13A
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human VPS13A.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23230
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.