Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS13B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127120 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-120 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-120 Supplier Abnova Corporation Supplier No. PAB25503
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VPS13B.

This gene encodes a potential transmembrane protein that may function in vesicle-mediated transport and sorting of proteins within the cell. This protein may play a role in the development and the function of the eye, hematological system, and central nervous system. Mutations in this gene have been associated with Cohen syndrome. Multiple splice variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Specifications

Antigen VPS13B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VPS13B.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene VPS13B
Gene Alias CHS1/COH1/DKFZp313I0811/KIAA0532
Gene Symbols VPS13B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human VPS13B.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157680
Test Specificity At least five alternatively spliced transcript variants have been observed. COH1 detects two isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.