Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS39, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-907 Supplier Abnova Corporation Supplier No. PAB19373
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of VPS39.

This gene encodes a protein that may promote clustering and fusion of late endosomes and lysosomes. The protein may also act as an adaptor protein that modulates the transforming growth factor-beta response by coupling the transforming growth factor-beta receptor complex to the Smad pathway. [provided by RefSeq

Specifications

Antigen VPS39
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of VPS39.
Dilution Western Blot (0.5 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene VPS39
Gene Alias FLJ21681/FLJ46546/KIAA0770/TLP/VAM6/hVam6p
Gene Symbols VPS39
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids near C-terminus of human VPS39.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23339
Test Specificity At least four isoforms of VPS39 are known to exist; this antibody will detect all four. VPS39 antibody is predicted not to cross-react with TRAP1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.