Learn More
WASF2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WASF2.
Supplier: Abnova Corporation PAB7889
Description
This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. The published map location (PMID:10381382) has been changed based on recent genomic sequence comparisons, which indicate that the expressed gene is located on chromosome 1, and a pseudogene may be located on chromosome X. [provided by RefSeq
Specifications
WASF2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of WASF2. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
SCAR2/WAVE2/dJ393P12.2 | |
Rabbit | |
100 μL | |
Primary | |
This sequence has high homology with similar regions in rat and mouse WAVE2, and has low homology to similar regions in WAVE1 and WAVE3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
WASF2 | |
WASF2 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human WASF2. | |
RUO | |
10163 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only