Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WASH6P, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WASH6P.
Supplier: Abnova Corporation PAB28212
Description
Sequence: GAPREVVDPSGGWATLLESIRQAGGIGKAKLRSMKERKLEKKKQKEQEQVRATSQGGHLMSDLFNKLVMRHKGISGKGPGAGEGPGGAFARVSDSIPPLPPPQQPQVDEDEDDWESSpecifications
WASH6P | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant WASH6P. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
CXYorf1/FAM39A/FLJ77647 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
653440 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/ml) Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
WASH6P | |
WASH6P | |
Recombinant protein corresponding to amino acids of recombinant WASH6P. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction