Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR45L, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122469 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-469 Supplier Abnova Corporation Supplier No. PAB20972

Rabbit polyclonal antibody raised against recombinant WDR45L.

This gene encodes a member of the WIPI or SVP1 family of WD40 repeat-containing proteins. The protein contains seven WD40 repeats that are thought to fold into a beta-propeller structure that mediates protein-protein interactions, and a conserved motif for interaction with phospholipids. The human genome contains several pseudogenes of this gene. [provided by RefSeq

Sequence: LFRCNYLALVGGGKKPKYPPNKVMIWDDLKKKTVIEIEFSTEVKAVKLRRDRIVVVLDSMIKVFTFTHNPHQLHVFETCYNPKGLCVLCPNSNNSLLAFPGTHTGHVQLVDLASTEKPPVDIPAHEGVLSCIALNLQGTRIATASEKG

Specifications

Antigen WDR45L
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description WDR45-like
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene WDR45L
Gene Alias WIPI-3, WIPI3
Gene Symbols WDR45L
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human WDR45L.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56270
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.