Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Core Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Core.
Supplier: Abnova Corporation PAB12942
Specifications
West Nile Virus Core | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Core. | |
In PBS (0.02% sodium azide) | |
A synthetic peptide corresponding to C-terminus 15 amino acids of West Nile Virus Core protein. | |
RUO | |
Virus | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction