Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whrn, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117848 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-848 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-848 Supplier Abnova Corporation Supplier No. PAB15785
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Whrn.

Specifications

Antigen Whrn
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Whrn.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Whrn
Gene Alias 1110035G07Rik/AW122018/AW742671/C430046P22Rik/MGC183668/bM340H1.8/mKIAA1526/wi
Gene Symbols Whrn
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 249 amino acids of mouse Whrn.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73750
Test Specificity Specific to recombinant protein GX0164. This antibody detects mWHRN protein. It also recognizes human WHRN protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.