Learn More
WIPI2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WIPI2.
Supplier: Abnova Corporation PAB21251
Description
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI2, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids (Proikas-Cezanne et al., 2004 [PubMed 15602573]).[supplied by OMIM
Sequence: ECALMKQHRLDGSLETTNEILDSASHDCPLVTQTYGAAAGKGTYVPSSPTRLAYTDDLGAVGGACLEDEASALRLDEDSEHPPMILRTDSpecifications
WIPI2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant WIPI2. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Atg21/CGI-50/DKFZp434J154/DKFZp686P02188/FLJ12979/FLJ14217/FLJ42984/WIPI-2 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
26100 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
WIPI2 | |
WIPI2 | |
Recombinant protein corresponding to amino acids of human WIPI2. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only