Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT11, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-854 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-854 Supplier Abnova Corporation Supplier No. PAB28352
Add to Cart
Add to Cart

Rabbit polyconal antibody raised against synthetic peptide of WNT11.

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 97%, 85%, and 63% amino acid identity with mouse, chicken, and Xenopus Wnt11 protein, respectively. This gene may play roles in the development of skeleton, kidney and lung, and is considered to be a plausible candidate gene for High Bone Mass Syndrome. [provided by RefSeq

Specifications

Antigen WNT11
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyconal antibody raised against synthetic peptide of WNT11.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene WNT11
Gene Alias HWNT11/MGC141946/MGC141948
Gene Symbols WNT11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at internal region of human WNT11.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7481
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Bat, Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.