Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT9A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89129877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-877 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-877 Supplier Abnova Corporation Supplier No. PAB28373
Add to Cart
Add to Cart

Rabbit polyconal antibody raised against synthetic peptide of WNT9A.

The WNT gene family consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is expressed in gastric cancer cell lines. The protein encoded by this gene shows 75% amino acid identity to chicken Wnt14, which has been shown to play a central role in initiating synovial joint formation in the chick limb. This gene is clustered with another family member, WNT3A, in the chromosome 1q42 region. [provided by RefSeq

Specifications

Antigen WNT9A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyconal antibody raised against synthetic peptide of WNT9A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene WNT9A
Gene Alias MGC138165/MGC141991/WNT14
Gene Symbols WNT9A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at internal region of human WNT9A.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7483
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except SCUBE2 (53%).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.