Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAF1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-368 Supplier Abnova Corporation Supplier No. PAB13116
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of XAF1.

X-linked inhibitor of apoptosis (XIAP; MIM 300079) is a potent member of the IAP family. All members of this family possess baculoviral IAP (BIR) repeats, cysteine-rich domains of approximately 80 amino acids that bind and inhibit caspases (e.g., CASP3; MIM 600636). XIAP has 3 BIR domains and a C-terminal RING zinc finger that possesses E3 ubiquitin ligase (see MIM 601623) activity. XAF1 antagonizes the anticaspase activity of XIAP and may be important in mediating apoptosis resistance in cancer cells (Liston et al., 2001 [PubMed 11175744]).[supplied by OMIM

Specifications

Antigen XAF1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of XAF1.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene XAF1
Gene Alias BIRC4BP/HSXIAPAF1/XIAPAF1
Gene Symbols XAF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human XAF1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54739
Test Specificity A 32 KDa band can be detected.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.