Learn More
XIAP Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of XIAP.
Supplier: Abnova Corporation PAB12923
Description
The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7. [provided by RefSeq
Specifications
XIAP | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of XIAP. | |
In PBS (0.02% sodium azide) | |
API3/BIRC4/ILP1/MIHA/XLP2 | |
Rabbit | |
100 μg | |
Primary | |
Anti-XIAP is human and mouse reactive. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
XIAP | |
XIAP | |
A synthetic peptide corresponding to C-terminus 13 amino acids of human XIAP. | |
RUO | |
331 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only