Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115928 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-928 Supplier Abnova Corporation Supplier No. PAB12581
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Xrcc5.

Sequence: LVYVQLPFMEDLRQYMFSSLKN

Specifications

Antigen XRCC5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Xrcc5.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Xrcc5
Gene Alias AI314015/Ku80/Ku86
Gene Symbols Xrcc5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 419-440 of Xrcc5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22596
Test Specificity Reacts with residues 419-440 [LVYVQLPFMEDLRQYMFSSLKN] of the 80 KDa Ku80 protein. Sequence is 100% conserved between human, mouse, rat and hamster.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.