Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YBX2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89126594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-594 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-594 Supplier Abnova Corporation Supplier No. PAB2492
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of YBX2.

YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins (Gu et al., 1998 [PubMed 9780336]; Tekur et al., 1999 [PubMed 10100484]).[supplied by OMIM

Specifications

Antigen YBX2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of YBX2.
Dilution Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene YBX2
Gene Alias CONTRIN/CSDA3/DBPC/MGC45104/MSY2
Gene Symbols YBX2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human YBX2.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51087
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.