Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YFP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant YFP.
Supplier: Abnova Corporation PAB17964
Specifications
YFP | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant YFP. | |
In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
Rabbit | |
100 μg | |
Primary | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) Immunoprecipitation (10-20 ug/mL) The optimal working dilution should be determined by the end user. | |
AAR85349 | |
Recombinant protein corresponding to YFP. | |
RUO | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction