Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YPEL1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127434 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-434 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-434 Supplier Abnova Corporation Supplier No. PAB25807
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of YPEL1.

This gene encodes a potential zinc-binding protein and is located in the region associated with DiGeorge syndrome on chromosome 22. The encoded protein may play a role in the regulation of cellular morphology and may function during craniofacial development. [provided by RefSeq

Specifications

Antigen YPEL1
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of YPEL1.
Dilution Western Blot (1 ug/mL) Immunocytochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene YPEL1
Gene Alias FKSG3/MGC64992
Gene Symbols YPEL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 11 amino acids at N-terminus of human YPEL1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29799
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.