Learn More
YWHAZ Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of YWHAZ.
Supplier: Abnova Corporation PAB27012
Description
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Several transcript variants that differ in the 5' UTR but that encode the same protein have been identified for this gene. [provided by RefSeq]
Specifications
YWHAZ | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Immunoprecipitor (1:50-1:200) The optimal working dilution should be determined by the end user. | |
YWHAZ | |
YWHAZ | |
A synthetic peptide corresponding to human YWHAZ. | |
100 μL | |
Primary | |
YWHAZ polyclonal antibody detects endogenous levels of YWHAZ protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of YWHAZ. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
KCIP-1/MGC111427/MGC126532/MGC138156 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
7534 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only