Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC17 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129550 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-550 Supplier Abnova Corporation Supplier No. PAB28032

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen ZCCHC17
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
Gene Symbols ZCCHC17
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ZCCHC17
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 51538
Target Species Human
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.