Learn More
ZEB1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of ZEB1.
Supplier: Abnova Corporation PAB19268
Description
ZEB1 encodes a zinc finger transcription factor that represses T-lymphocyte-specific IL2 gene (MIM 147680) expression by binding to a negative regulatory domain 100 nucleotides 5-prime of the IL2 transcription start site (Williams et al., 1991 [PubMed 1840704]).[supplied by OMIM
Specifications
ZEB1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunocytochemistry (20 ug/mL) The optimal working dilution should be determined by the end user. | |
ZEB1 | |
ZEB1 | |
A synthetic peptide corresponding to 15 amino acids near internal region of human ZEB1. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of ZEB1. | |
In PBS (0.02% sodium azide) | |
AREB6/BZP/DELTA-EF1/MGC133261/NIL-2-A/NIL-2A/NIL2A/TCF8/ZEB/ZFHEP/ZFHX1A | |
Rabbit | |
Peptide affinity purification | |
RUO | |
6935 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only