Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zfp354c, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-920 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-920 Supplier Abnova Corporation Supplier No. PAB15858
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Zfp354c.

Specifications

Antigen Zfp354c
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Zfp354c.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Zfp354c
Gene Alias 5330421P20Rik/AJ18/AW743325/KIAA4218/Kid3/Tcf17l2/mKIAA4218
Gene Symbols Zfp354c
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 219 amino acids of mouse Zfp354c.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30944
Test Specificity Specific to recombinant protein GX0838. This antibody detects mZNF354C protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.