Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFPM2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-762 Supplier Abnova Corporation Supplier No. PAB20259
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ZFPM2.

The zinc finger protein encoded by this gene is a widely expressed member of the FOG family of transcription factors. The family members modulate the activity of GATA family proteins, which are important regulators of hematopoiesis and cardiogenesis in mammals. It has been demonstrated that the protein can both activate and down-regulate expression of GATA-target genes, suggesting different modulation in different promoter contexts. A related mRNA suggests an alternatively spliced product but this information is not yet fully supported by the sequence. [provided by RefSeq

Sequence: CLPEQEQRPPLVQQRFLDVANLNNPCTSTQEPTEGLGECYHPRCDIFPGIVSKHLETSLTINKCVPVSKCDTTHSSVSCLEMDVPIDLSKKCLSQSERTTTSPKRLLDYHECTVCKISFNKVENYLAHKQ

Specifications

Antigen ZFPM2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ZFPM2.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ZFPM2
Gene Alias DIH3/FOG2/MGC129663/MGC129664/ZNF89B/hFOG-2
Gene Symbols ZFPM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ZFPM2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23414
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.