Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zfyve16, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117823 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-823 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-823 Supplier Abnova Corporation Supplier No. PAB15759
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Zfyve16.

Specifications

Antigen Zfyve16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Zfyve16.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Zfyve16
Gene Alias AI035632/B130024H06Rik/B130031L15/mKIAA0305
Gene Symbols Zfyve16
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 153 amino acids of mouse Zfyve16.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218441
Test Specificity Specific to recombinant protein GX0350. This antibody detects mZFYVE16 protein. It also recognizes human ZFYVE16 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.