Learn More
ZFYVE26, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZFYVE26.
Supplier: Abnova Corporation PAB16701
Description
This gene encodes a protein which contains a FYVE zinc finger binding domain. The presence of this domain is thought to target these proteins to membrane lipids through interaction with phospholipids in the membrane. Mutations in this gene are associated with autosomal recessive spastic paraplegia-15. [provided by RefSeq
Specifications
ZFYVE26 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ZFYVE26. | |
In PBS (0.02% sodium azide) | |
DKFZp686F19106/DKFZp781H1112/KIAA0321/SPG15 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
ZFYVE26 | |
ZFYVE26 | |
A synthetic peptide corresponding C-terminus to 16 amino acids of human ZFYVE26. | |
RUO | |
23503 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only