Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMIZ2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89120900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-900 Supplier Abnova Corporation Supplier No. PAB19367
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ZMIZ2.

ZMIZ2 and ZMIZ1 (MIM 607159) are members of a PIAS (see MIM 603566)-like family of proteins that interact with nuclear hormone receptors. ZMIZ2 interacts with androgen receptor (AR; MIM 313700) and enhances AR-mediated transcription (Huang et al., 2005 [PubMed 16051670]).[supplied by OMIM

Specifications

Antigen ZMIZ2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ZMIZ2.
Dilution Western blot (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ZMIZ2
Gene Alias DKFZp761I2123/KIAA1886/ZIMP7/hZIMP7
Gene Symbols ZMIZ2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids near the center region of human ZMIZ2.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83637
Test Specificity At least four isoforms are known to exist. This antibody is predicted to not cross-react with other PIAS protein family members.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.