Learn More
ZMPSTE24 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
Supplier: Abnova Corporation PAB4317
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq
Specifications
ZMPSTE24 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24. | |
In PBS (0.09% sodium azide) | |
FACE-1/FACE1/FLJ14968/STE24/Ste24p | |
Rabbit | |
Protein G purification | |
RUO | |
10269 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
ZMPSTE24 | |
ZMPSTE24 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human ZMPSTE24. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only