Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF10, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-232 Supplier Abnova Corporation Supplier No. PAB28716

Rabbit polyclonal antibody raised against recombinant ZNF10.

The protein encoded by this gene contains a C2H2 zinc finger, and has been shown to function as a transcriptional repressor. The Kruppel-associated box (KRAB) domain of this protein is found to be responsible for its transcriptional repression activity. RING finger containing protein TIF1 was reported to interact with the KRAB domain, and may serve as a mediator for the repression activity of this protein. [provided by RefSeq

Sequence: RLEKGEEPWLVEREIHQETHPDSETAFEIKSSVSSRSIFKDKQSCDIKMEGMARNDLWYLSLEEVWKCRDQLDKYQENPERHLRQVAFTQKKVLTQERVSESGKYGGNCLLPAQLV

Specifications

Antigen ZNF10
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ZNF10.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene ZNF10
Gene Alias KOX1
Gene Symbols ZNF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ZNF10.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7556
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.