Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF160, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123091 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-091 Supplier Abnova Corporation Supplier No. PAB21594
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ZNF160.

The protein encoded by this gene is a Kruppel-related zinc finger protein which is characterized by the presence of an N-terminal repressor domain, the Kruppel-associated box (KRAB). The KRAB domain is a potent repressor of transcription; thus this protein may function in transcription regulation. Three alternative transcripts encoding the same protein have been described. [provided by RefSeq

Sequence: LVSLGLCHFDMNIISMLEEGKEPWTVKSCVKIARKPRTRECVKGVVTDIPPKCTIKDLLPKEKSSTEAVFHTVVLERHESPDIEDFSFKEPQKNVHDFECQWRDDTGNYKGVLMAQKEGKRDQRDRRDIENKLMNNQLGVSFHSHLP

Specifications

Antigen ZNF160
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ZNF160.
Dilution Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ZNF160
Gene Alias DKFZp686B16128/F11/FLJ00032/HKr18/HZF5/KIAA1611/KR18
Gene Symbols ZNF160
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ZNF160.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90338
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.