Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF295, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZNF295.
Supplier: Abnova Corporation PAB21732
Description
Sequence: QPEPNKVNHIVTTKDDNVFSDSSEQVNFDSEDSSCLPEDLSLSKQLKIQVKEEPVEEAEEEAPEASTAPKEAGPSKEASLWPCEKCGKMFTVHKQLERHQELLCSVKPFICHVCNKAFRTNFRLWSHFQSHMSQASEESAHKESEVCPSpecifications
ZNF295 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ZNF295. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
DKFZp781N1974/ZBTB21 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
49854 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
ZNF295 | |
ZNF295 | |
Recombinant protein corresponding to amino acids of human ZNF295. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction