Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zscan12, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117783 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-783 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-783 Supplier Abnova Corporation Supplier No. PAB15718
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Zscan12.

Sequence: PGRKVHGCDECGKSFTQHSRLIEHKRVHTGDRPYKCEVCGKTFRWRTVLIRHKVVHTGEKPYKCNECGRAFGQWSALNQHQRLHSGEKHYHCNECGKAFCQKAGLFHHLKSHRRNRPYQCLQCNKSFNRRSTLSQHQGVHTGAKPYECNDCGKAFVYNSSLATHQETHHKEKPFTQSGPIQQQRNHTKEKPYKCSVCGKAFIQKISLIEHEQIHTGERPYKCAEGGKAFIQMSELTEH

Specifications

Antigen Zscan12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Zscan12.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Zscan12
Gene Alias 2510038J07Rik/FPM315/Zfp96/mKIAA0426
Gene Symbols Zscan12
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 238 mouse Zscan12.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22758
Test Specificity Specific to recombinant protein GX0926. This antibody detects mZSCAN12.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.