Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123378 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-378 Supplier Abnova Corporation Supplier No. PAB21681
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ZSCAN2.

The protein encoded by this gene contains several copies of zinc finger motif, which is commonly found in transcriptional regulatory proteins. Studies in mice show that this gene is expressed during embryonic development, and specifically in the testis in adult mice, suggesting that it may play a role in regulating genes in germ cells. Alternative splicing of this gene results in several transcript variants encoding different isoforms. [provided by RefSeq

Sequence: ATHRRTHMVEKPYKCGVCGKSFSQSSSLIAHQGMHTGEKPYECLTCGESFSWSSNLLKHQRIHTGEKPYKCSECGKCFSQRSQLVVHQRTHTGEKPYKCLMCGKSFSRGSILVMHQRAHLGDKPYRCPECGKGFSWNSVLII

Specifications

Antigen ZSCAN2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ZSCAN2.
Dilution Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene ZSCAN2
Gene Alias FLJ20595/ZFP29/ZNF854
Gene Symbols ZSCAN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ZSCAN2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54993
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.