Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Fc Goat anti-Rabbit, Clone: RMG02 (Biotin), Abnova™

Catalog No. 89035296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-296 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-296 Supplier Abnova Corporation Supplier No. MAB14944
Add to Cart
Add to Cart

Goat Monoclonal Antibody

Goat monoclonal antibody raised against native rabbit IgG Fc.

  • High specificity
  • High reactivity

Specifications

Antigen Rabbit IgG Fc
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RMG02
Conjugate Biotin
Description Goat monoclonal antibody raised against native rabbit IgG Fc.
Dilution ELISA (0.01-0.5 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Host Species Goat
Immunogen Native purified rabbit IgG.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rabbit
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.