Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Heavy chain (Fc fragment) Monoclonal Antibody (A6-B8), Huabio

Catalog No. 502544493
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4493 Supplier HUABIO RESEARCH INC Supplier No. M09025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rabbit IgG Heavy chain (Fc fragment) Monoclonal antibody specifically detects Rabbit IgG Heavy chain (Fc fragment) in Rabbit samples. It is validated for ELISA, Western Blot

Specifications

Antigen Rabbit IgG Heavy chain (Fc fragment)
Applications ELISA, Western Blot
Classification Monoclonal
Clone A6-B8
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 25% glycerol and 0.05% sodium azide; pH 7.4
Host Species Mouse
Immunogen Purified Rabbit IgG heavy chain (Fc fragment).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rabbit
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.