Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Isotype Control, PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB398393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398393 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398393 Supplier Novus Biologicals Supplier No. NBP236463PECY7
Add to Cart
Add to Cart

Rabbit Isotype Control

IgG Isotype Control specifically detects IgG in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page, Flow (Intracellular), ChIP assay (ChIP)

Specifications

Antigen IgG
Applications Western Blot, ELISA, Immunohistochemistry, SDS-Page, Radioimmune Assays (RIA)
Conjugate PE-Cyanine7
Formulation PBS
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Gene ID (Entrez) 3500
Target Species Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.