Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit IL-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6939
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Rabbit samples. It is validated for Neutralization.Specifications
IL-4 | |
Monoclonal | |
LYOPH | |
Neutralization | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects rabbit IL-4 in ELISAs. | |
Rabbit | |
Purified |
Neutralization | |
855824 | |
Unconjugated | |
Q9MZR8 | |
IL4 | |
E. coli-derived recombinant rabbit IL-4 Arg25-Ser147 Accession # Q9MZR8 | |
100 μg | |
3565 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction