Learn More
Description
Specifications
Specifications
| Assay Range | Request Information |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum, Tissue Homogenates |
| Target Species | Rabbit |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Research Discipline | Signal Transduction |
| Sample Volume | 50 to 100 μL |
| Storage Requirements | Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
| Synonym | T4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
