Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB56701
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB56701 100 μg
MAB56701SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB56701 Supplier R&D Systems Supplier No. MAB56701
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Rabbit samples. It is validated for Western Blot.

Specifications

Antigen TNF-alpha
Applications Western Blot
Classification Monoclonal
Clone 610325
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04924
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen E. coli-derived recombinant rabbit TNF-alpha Leu79-Leu234 Accession # P04924
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects rabbit TNF-alpha in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant cotton rat, feline, guinea pig, human, mouse, rat TNF-alpha , less than 10% cross-reactivity with recombinant canine, equine, rhesus TNF-alpha , and no cross-reactivity with recombinant bovine or porcine TNF-alpha is observed. No cross-reactivity with recombinant human (rh) 4-1BB Ligand, APRIL, rhBAFF, rhEDA, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1 beta 2, rhLymphotoxin alpha 2 beta 1, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, rhVEGI, recombinant mouse (rm) 4-1BB Ligand, rmBAFF, rmFas Ligand, rmGITR Ligand, rmLIGHT, rmOX40 Ligand, rmTRAIL, rmTRAIL-2 Ligand, rmTRANCE, or rmTWEAK is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Rabbit
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.