Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabenosyn 5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733777 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3777 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3777 Supplier Proteintech Group Inc Supplier No. 222181AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.

Specifications

Antigen Rabenosyn 5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBSN
Gene Accession No. Q80Y56, Q9H1K0
Gene Alias 110 kDa protein, Rabenosyn 5, ZFYVE20
Gene Symbols RBSN
Host Species Rabbit
Immunogen Rabenosyn 5 Fusion Protein Ag17554
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64145, 78287
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.