Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabenosyn 5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30333220UL
Description
Rabenosyn 5 Polyclonal specifically detects Rabenosyn 5 in Mouse samples. It is validated for Western Blot.Specifications
Rabenosyn 5 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
64145 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
110 kDa protein, FLJ34993, FYVE finger-containing Rab5 effector protein rabenosyn-5, FYVE-finger-containing Rab5 effector protein rabenosyn-5, MGC126210, Rabenosyn-5, ZFYVE20, Zinc finger FYVE domain-containing protein 20, zinc finger, FYVE domain containing 20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Rabenosyn 5 (NP_071735.2). | |
20 μL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction