Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RABEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118232 Supplier Invitrogen™ Supplier No. PA5118232
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RABEP1 Polyclonal Antibody for Western Blot, ICC/IF, IP

RABEP1 gene ontology annotations related to this gene include endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABEP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene RABEP1
Gene Accession No. Q15276
Gene Alias neurocrescin; rab GTPase-binding effector protein 1; RAB5 effector protein; RAB5EP; rabaptin 5; rabaptin, RAB GTPase binding effector protein 1; rabaptin-4; Rabaptin-5; rabaptin-5alpha; Rabep1; RABPT5; Rabpt5a; Renal carcinoma antigen NY-REN-17
Gene Symbols RABEP1
Host Species Rabbit
Immunogen E. coli-derived Human RABEP1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.