Learn More
RABEP1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302821A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
RABEP1 gene ontology annotations related to this gene include endocytosis.Specifications
RABEP1 | |
Polyclonal | |
Unconjugated | |
RABEP1 | |
Rabaptin-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54189, 9135 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O35551, Q15276 | |
RABEP1 | |
Between 812 and 862 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.