Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RABEP2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306404PE
Description
RABEP2 Polyclonal antibody specifically detects RABEP2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
RABEP2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
FLJ23282, FRA, rab GTPase-binding effector protein 2, rabaptin, RAB GTPase binding effector protein 2, Rabaptin-5 beta, rabaptin-5beta, RABPT5B | |
Produced in rabbits immunized with E. coli-derived Human RABEP2 fragment. | |
0.1 mL | |
Cytokine Research | |
79874 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction